Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027187.3 | NP_081463.1 | 4 | 413 | ||
GenBank mRNA | AK146518.1 | - | 4 | 414 | ||
BC029252.1 | - | 4 | 443 | 3666709 | ||
BC038158.1 | - | 4 | 487 | 1493985 |