Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145954.1 | NP_666066.1 | 15 | 2133 | ||
GenBank mRNA | AK010387.1 | - | 2 | 236 | ||
AK030673.1 | - | 15 | 2126 | |||
AK144967.1 | - | 2132 | ||||
AK144995.1 | - | 15 | 2140 | |||
AK151691.1 | - | 15 | 2133 | |||
AK153129.1 | - | 8 | 1040 | |||
AK161198.1 | - | 15 | 2171 | |||
AK164374.1 | - | 15 | 2179 | |||
AK169346.1 | - | 2484 | ||||
AK170276.1 | - | 15 | 2172 | |||
AK182316.1 | - | 1 | 71 | |||
AK220200.1 | - | 15 | 2109 | |||
BC013548.1 | - | 15 | 2133 | 4222058 | ||
BC030467.1 | - | 2 | 341 |