Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005934.3 | NP_005925.2 | 12 | 2803 | ||
XM_011528021.1 | XP_011526323.1 | 12 | 2777 | |||
XM_011528022.1 | XP_011526324.1 | 12 | 2660 | |||
XM_011528023.1 | XP_011526325.1 | 11 | 2651 | |||
XM_017026819.1 | XP_016882308.1 | 12 | 3359 | |||
GenBank mRNA | AL365410.1 | - | 1 | 148 | ||
BC047008.1 | - | 1 | 338 | |||
BC084564.1 | - | 1 | 309 |