Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133716.3 | NP_598477.2 | 7 | 991 | ||
GenBank mRNA | AK152757.1 | - | 7 | 958 | ||
BC031581.1 | - | 7 | 980 | |||
BC032217.1 | - | 7 | 967 | |||
BC052413.1 | - | 7 | 991 | 5720177 |