Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027215.2 | NP_081491.2 | 1 | 162 | ||
GenBank mRNA | AK007753.1 | - | 1 | 123 | ||
AK008089.1 | - | 1 | 116 | |||
AK010168.1 | - | 1 | 151 | |||
AK160267.1 | - | 1 | 164 | |||
BC035323.1 | - | 1 | 135 | |||
BC147300.1 | - | 1 | 116 | 9056333 | ||
BC147301.1 | - | 1 | 116 | 9056336 |