Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027216.5 | NP_081492.4 | 5, 6 | 574 | ||
GenBank mRNA | AK007986.1 | - | 5, 6 | 514 | ||
AK036999.1 | - | 5, 6 | 521 | |||
AK170673.1 | - | 5, 6 | 514 | |||
BC004643.1 | - | 1, 2 | 104 | |||
BC019647.1 | - | 5, 6 | 518 | 3978277 | ||
BC031751.1 | - | 6, 7 | 632 | 4216487 |