Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190310.1 | NP_001177239.1 | 6 | 679 | ||
NM_027218.3 | NP_081494.1 | 5 | 580 | |||
GenBank mRNA | AK007794.1 | - | 5 | 537 | ||
AY230259.1 | - | 6 | 643 | |||
AY230260.1 | - | 5 | 544 | |||
AY365134.1 | - | 6 | 575 |