Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtcap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027221.3 | NP_081497.2 | 4 | 463 | ||
GenBank mRNA | AK007999.1 | - | 4 | 376 | ||
BC016516.1 | - | 3 | 573 | 2609442 | ||
BC027812.1 | - | 4 | 461 | 5251276 | ||
BC107024.1 | - | 4 | 440 | 40049838 | ||
BC107025.1 | - | 4 | 440 | 40049842 | ||
CK129285.1 | - | 4 | 463 |