Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198834.2 | NP_942131.1 | 23 | 3490 | ||
NM_198836.2 | NP_942133.1 | 23 | 3058 | |||
NM_198837.1 | NP_942134.1 | 21 | 3128 | |||
NM_198838.1 | NP_942135.1 | 22 | 3239 | |||
NM_198839.2 | NP_942136.1 | 27 | 4164 | |||
XM_005257267.4 | XP_005257324.1 | 23 | 2971 | |||
XM_006721853.1 | XP_006721916.1 | 23 | 3501 | |||
XM_011524701.1 | XP_011523003.1 | 23 | 3411 | |||
XM_011524703.1 | XP_011523005.1 | 23 | 3043 | |||
XM_011524704.2 | XP_011523006.1 | 23 | 2976 | |||
XM_017024553.1 | XP_016880042.1 | 24 | 3525 | |||
XM_017024554.1 | XP_016880043.1 | 24 | 3525 | |||
XM_017024555.1 | XP_016880044.1 | 23 | 3124 | |||
GenBank mRNA | AB371587.1 | - | 23 | 3037 | ||
AK309084.1 | - | 23 | 3046 | |||
AY237919.1 | - | 21 | 2867 | |||
AY315627.1 | - | 21 | 2867 | |||
BC137287.1 | - | 23 | 2995 | 9021284 | ||
U19822.1 | - | 21 | 2867 |