Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1cf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081074.1 | NP_001074543.1 | 6 | 899 | ||
GenBank mRNA | AK007970.1 | - | 5 | 743 | ||
AY566862.1 | - | 6 | 896 | |||
AY566863.1 | - | 6 | 896 | |||
AY566864.1 | - | 6 | 896 | |||
BC038942.1 | - | 4 | 599 | |||
BC089622.1 | - | 6 | 847 | 6760455 |