Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077631.2 | NP_001071099.1 | 5 | 914 | ||
NM_175153.4 | NP_780362.2 | 5 | 1135 | |||
GenBank mRNA | AK032095.1 | - | 5 | 1129 | ||
AK035654.1 | - | 5 | 842 | |||
AK041578.1 | - | 5 | 830 | |||
AK050654.1 | - | 5 | 1121 | |||
AK086850.1 | - | 5 | 828 | |||
AK168222.1 | - | 4 | 817 | |||
AK169772.1 | - | 5 | 874 | |||
BC024913.1 | - | 4 | 819 | 3602181 |
Gene Symbol: 6330549D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032024.1 | - | 2 | 1161 | ||
AK088233.1 | - | 3 | 1561 |