Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077631.2 | NP_001071099.1 | 8 | 1298 | ||
NM_175153.4 | NP_780362.2 | 8 | 1519 | |||
GenBank mRNA | AK032095.1 | - | 8 | 1513 | ||
AK035654.1 | - | 8 | 1226 | |||
AK050654.1 | - | 8 | 1505 | |||
AK086850.1 | - | 8 | 1212 | |||
AK168222.1 | - | 7 | 1201 | |||
AK169772.1 | - | 8 | 1258 | |||
BC024913.1 | - | 7 | 1203 | 3602181 |
Gene Symbol: 6330549D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032024.1 | - | 2 | 1533 | ||
AK088233.1 | - | 3 | 1933 | |||
BC147330.1 | - | 1 | 356 | |||
BC147331.1 | - | 1 | 356 |