Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6330549D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032024.1 | - | 2 | 825 | ||
AK088233.1 | - | 3 | 1225 |
Gene Symbol: Vwa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077631.2 | NP_001071099.1 | 3 | 577 | ||
NM_175153.4 | NP_780362.2 | 3 | 798 | |||
GenBank mRNA | AK032095.1 | - | 3 | 792 | ||
AK035654.1 | - | 3 | 505 | |||
AK041578.1 | - | 3 | 493 | |||
AK050654.1 | - | 3 | 784 | |||
AK086850.1 | - | 3 | 490 | |||
AK168222.1 | - | 2 | 480 | |||
AK169772.1 | - | 3 | 537 | |||
BC024913.1 | - | 2 | 482 | 3602181 |