Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011707.1 | NP_001011707.1 | 4 | 615 | ||
GenBank mRNA | AK008512.2 | - | 3 | 441 | ||
AY227734.1 | - | 4 | 615 | |||
BC147819.1 | - | 4 | 612 | 9087536 | ||
BC147824.1 | - | 4 | 612 | 9087541 |