Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290810.1 | NP_001277739.1 | 8 | 884 | ||
NM_023536.3 | NP_076025.1 | 8 | 881 | |||
GenBank mRNA | AK011387.1 | - | 8 | 606 | ||
AK171445.1 | - | 2 | 2769 | |||
AK186912.1 | - | 2 | 65 | |||
BC005734.1 | - | 8 | 603 | 3598883 | ||
BC096556.1 | - | 8 | 627 | |||
BC138221.1 | - | 8 | 683 | 8861240 | ||
BC145205.1 | - | 8 | 683 | 9053756 | ||
BC145209.1 | - | 8 | 705 |