Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027268.2 | NP_081544.1 | 6, 7 | 1057 | ||
GenBank mRNA | AK012765.1 | - | 6, 7 | 1085 | ||
AK032573.1 | - | 6, 7 | 1084 | |||
AK050481.1 | - | 6, 7 | 1075 | |||
AK082953.1 | - | 6, 7 | 1057 | |||
AK089968.1 | - | 6, 7 | 1047 | |||
AK129084.1 | - | 6, 7 | 1044 | |||
AK145904.1 | - | 6, 7 | 1059 | |||
BC023889.1 | - | 6, 7 | 1044 | 5252046 | ||
BC058619.1 | - | 1 | 2596 |