Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810021J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172403.2 | NP_765991.1 | 5 | 747 | ||
GenBank mRNA | AK030099.1 | - | 5 | 772 | ||
AK134479.1 | - | 5 | 772 | |||
BC058081.1 | - | 5 | 747 | 6830592 |