Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027275.3 | NP_081551.2 | 7 | 477 | ||
GenBank mRNA | AK032831.1 | - | 7 | 461 | ||
AK146005.1 | - | 7 | 466 | |||
AK146172.1 | - | 7 | 478 | |||
AK172480.1 | - | 7 | 466 | |||
BC115434.2 | - | 7 | 473 | 40094203 |