Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027276.2 | NP_081552.2 | 13 | 1238 | ||
GenBank mRNA | AK013213.1 | - | 2 | 172 | ||
AK147938.1 | - | 13 | 1253 | |||
AK167965.1 | - | 13 | 1253 | |||
AK168978.1 | - | 13 | 1225 | |||
AK190066.1 | - | 2 | 139 | |||
AK213282.1 | - | 2 | 139 | |||
AK215012.1 | - | 2 | 139 | |||
BC026606.1 | - | 13 | 1238 | 4976424 |