Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081137.1 | NP_001074606.1 | 46, 47 | 5385 | ||
GenBank mRNA | AK008441.1 | - | 3, 4 | 264 | ||
EU937530.1 | - | 47, 48 | 5448 |