Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023544.5 | NP_076033.4 | 1 | 262 | ||
GenBank mRNA | AI854132.1 | - | 1 | 309 | ||
BC150752.1 | - | 1 | 262 | 9087663 |
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017966.2 | NP_001017966.1 | 10 | 1647 | ||
GenBank mRNA | AK129015.1 | - | 1626 | |||
BC021415.1 | - | 10 | 1628 | |||
BC031491.1 | - | 3 | 401 |