Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144527.3 | NP_653110.3 | 14 | 2308 | ||
GenBank mRNA | AK030742.1 | - | 14 | 2273 | ||
AK141103.1 | - | 14 | 2298 | |||
AK167665.1 | - | 3 | 304 |