Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023547.1 | NP_076036.1 | 3 | 307 | ||
GenBank mRNA | AK010946.1 | - | 3 | 409 | ||
BC019383.1 | - | 3 | 341 | 5010975 | ||
BC047990.1 | - | 3 | 342 | |||
BC058788.1 | - | 3 | 353 | 6440887 |