Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145938.1 | NP_001139410.1 | 2 | 203 | ||
NM_002421.3 | NP_002412.1 | 2 | 355 | |||
GenBank mRNA | AK024818.1 | - | 2 | 278 | ||
AK130687.1 | - | 2 | 278 | |||
AK223035.1 | - | 2 | 236 | |||
AK223056.1 | - | 1 | 21 | |||
AK223281.1 | - | 2 | 278 | |||
AK225495.1 | - | 1 | 46 | |||
AK297723.1 | - | 2 | 126 | |||
AK301340.1 | - | 2 | 281 | |||
BC013875.2 | - | 2 | 254 | 3834572 | ||
BT006874.1 | - | 2 | 212 | |||
BT020147.1 | - | 2 | 212 | |||
DC385245.1 | - | 2 | 355 | |||
M13509.1 | - | 2 | 280 | |||
X05231.1 | - | 2 | 278 | |||
X54925.1 | - | 2 | 280 |