Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197993.2 | NP_932110.1 | 5 | 1167 | ||
GenBank mRNA | AK138418.1 | - | 5 | 1087 | ||
BC023900.1 | - | 5 | 1144 | 5321265 | ||
BC025499.1 | - | 5 | 1191 | 5320398 |