Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027308.2 | NP_081584.1 | 2 | 413 | ||
GenBank mRNA | AK008696.1 | - | 2 | 415 | ||
AK033677.1 | - | 2 | 414 | |||
AK088927.1 | - | 3 | 369 | |||
AK135657.1 | - | 219 | ||||
BC139455.1 | - | 2 | 261 | 8862477 | ||
BC139458.1 | - | 2 | 261 | 8862480 |