Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175155.4 | NP_780364.3 | 9 | 989 | ||
GenBank mRNA | AJ507736.1 | - | 9 | 804 | ||
AK129216.1 | - | 4 | 323 | |||
BY770549.1 | - | 2 | 211 |