Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144801.2 | NP_659050.2 | 7 | 1027 | ||
GenBank mRNA | AK035836.1 | - | 6 | 884 | ||
AK039173.1 | - | 6 | 1097 | |||
AK087636.1 | - | 6 | 1232 | |||
BC017618.1 | - | 7 | 1021 | 4923610 |
Gene Symbol: 2900057H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013719.1 | - | 666 |