Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142950.1 | NP_001136422.1 | 2 | 199 | ||
NM_027350.3 | NP_081626.2 | 2 | 196 | |||
GenBank mRNA | AK077699.1 | - | 2 | 177 | ||
AK083730.1 | - | 2 | 156 | |||
AK157244.1 | - | 2 | 158 | |||
AK172657.1 | - | 2 | 158 | |||
AK191424.1 | - | 1 | 17 | |||
BC005473.1 | - | 2 | 102 | |||
BC052849.1 | - | 2 | 157 | 30057822 | ||
BY117114.1 | - | 2 | 200 |