Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027354.2 | NP_081630.2 | 3, 4 | 656 | ||
GenBank mRNA | AK011064.1 | - | 3, 4 | 334 | ||
AK154163.1 | - | 3, 4 | 406 | |||
BC034901.1 | - | 3, 4 | 316 | 5255069 | ||
CA557124.1 | - | 3, 4 | 306 |