Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhlhb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098222.1 | NP_001091692.1 | 5 | 1586 | ||
NM_198161.2 | NP_937804.1 | 5 | 1596 | |||
GenBank mRNA | BC059871.1 | - | 1 | 951 | 6809364 |