Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027373.2 | NP_081649.1 | 5 | 583 | ||
GenBank mRNA | AK011153.1 | - | 5 | 411 | ||
AK080756.1 | - | 5 | 518 | |||
AK129483.1 | - | 5 | 510 | |||
AK147706.1 | - | 5 | 475 | |||
AK148000.1 | - | 5 | 438 | |||
AK161385.1 | - | 5 | 534 | |||
BC050814.1 | - | 5 | 583 | 6773872 |