Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030566.2 | NP_085043.2 | 9 | 1656 | ||
GenBank mRNA | AK029188.1 | - | 7 | 1014 | ||
AK089938.1 | - | 9 | 1361 | |||
AK144836.1 | - | 9 | 1342 | |||
AK157832.1 | - | 9 | 1317 | |||
BC004088.1 | - | 8 | 1402 | 3593722 | ||
BC015287.1 | - | 9 | 1664 | 4240442 |