Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197995.2 | NP_932112.2 | 3 | 380 | ||
GenBank mRNA | AK011161.1 | - | 2 | 176 | ||
AK038349.1 | - | 3 | 252 | |||
AK140077.1 | - | 2 | 384 | |||
BC009089.1 | - | 3 | 221 | |||
BC139260.1 | - | 3 | 251 | 8862282 | ||
BC139262.1 | - | 3 | 251 | 8862284 | ||
BF472168.1 | - | 3 | 387 |