Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077636.1 | NP_001071104.1 | 2 | 1382 | ||
NM_027388.2 | NP_081664.2 | 2 | 1801 | |||
GenBank mRNA | AK081337.1 | - | 2 | 1802 | ||
AK086537.1 | - | 2 | 1383 | |||
BC125492.1 | - | 2 | 1735 | |||
BC144942.1 | - | 2 | 1735 | 9053490 |