Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177608.3 | NP_808276.2 | 12 | 1903 | ||
GenBank mRNA | AK049924.1 | - | 12 | 1906 | ||
AK145135.1 | - | 12 | 1789 | |||
AK169713.1 | - | 12 | 1790 | |||
AK172920.1 | - | 12 | 1770 | |||
BC071239.1 | - | 5 | 531 | |||
BC095939.1 | - | 11 | 1866 | |||
BC141056.1 | - | 12 | 1863 | 9055668 | ||
CB528069.1 | - | 2 | 133 | |||
CJ198024.1 | - | 3 | 405 |