Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133726.2 | NP_598487.1 | 12 | 1169 | ||
GenBank mRNA | AK013981.1 | - | 12 | 1152 | ||
AK146528.1 | - | 12 | 1165 | |||
AK147169.1 | - | 1192 | ||||
AK150686.1 | - | 1140 | ||||
AK151631.1 | - | 1140 | ||||
AK152270.1 | - | 12 | 1165 | |||
AK154658.1 | - | 1158 | ||||
AK159149.1 | - | 1149 | ||||
AK168557.1 | - | 7 | 691 | |||
AK205875.1 | - | 1 | 70 | |||
BC003843.1 | - | 12 | 1169 | 3495262 | ||
CT010318.1 | - | 12 | 1069 |