Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190885.1 | NP_001177814.1 | 6 | 776 | ||
NM_001190886.1 | NP_001177815.1 | 5 | 732 | |||
NM_001290690.1 | NP_001277619.1 | 5 | 931 | |||
NM_027398.3 | NP_081674.2 | 5 | 743 | |||
GenBank mRNA | AB075041.1 | - | 5 | 373 | ||
AJ278534.1 | - | 4 | 301 | |||
AK014313.1 | - | 5 | 732 | |||
AK045948.1 | - | 6 | 777 | |||
AK046398.1 | - | 6 | 765 | |||
AK048178.1 | - | 3 | 2756 | |||
AK081845.1 | - | 5 | 733 | |||
AK082267.1 | - | 4 | 1462 | |||
AY050525.1 | - | 6 | 381 | |||
AY050526.1 | - | 5 | 348 | |||
AY171234.1 | - | 5 | 807 | |||
AY647242.1 | - | 5 | 575 | |||
BC034241.1 | - | 5 | 685 | 4503251 | ||
BU702946.1 | - | 4 | 429 | |||
DQ148493.1 | - | 5 | 685 | |||
DQ148494.1 | - | 5 | 733 | |||
DQ148495.1 | - | 6 | 381 |