Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032544.3 | NP_115933.2 | 2 | 791 | ||
GenBank mRNA | AF361482.2 | - | 2 | 791 | ||
AK046330.1 | - | 2 | 167 | |||
AK204819.1 | - | 2 | 72 | |||
BC055716.1 | - | 2 | 143 | |||
BC075712.1 | - | 2 | 151 | |||
BC092042.1 | - | 2 | 174 | |||
BC138560.1 | - | 2 | 200 | 8861580 | ||
BC145333.1 | - | 2 | 200 |