Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290514.1 | NP_001277443.1 | 5 | 801 | ||
NM_027409.5 | NP_081685.1 | 5 | 804 | |||
GenBank mRNA | AK007533.1 | - | 5 | 737 | ||
AK088275.1 | - | 5 | 738 | |||
AK139795.1 | - | 5 | 745 | |||
AK203602.1 | - | 1 | 52 | |||
BC018329.1 | - | 5 | 702 | 3962906 | ||
BC085100.1 | - | 5 | 709 | 6391017 | ||
BC100442.1 | - | 5 | 689 | 30501889 | ||
DV651027.1 | - | 2 | 171 |