Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027411.2 | NP_081687.2 | 9, 10 | 1309 | ||
GenBank mRNA | AK139553.1 | - | 9, 10 | 1185 | ||
BC006674.1 | - | 9, 10 | 1292 | 3499487 |