Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002428.3 | NP_002419.1 | 8 | 2109 | ||
GenBank mRNA | BC006412.1 | - | 8 | 1229 | ||
BC036495.1 | - | 8 | 2115 | 5263145 | ||
BC055428.1 | - | 8 | 2110 | 4811572 | ||
BT006715.1 | - | 6 | 1009 | |||
D85510.1 | - | 5 | 839 | |||
D86331.1 | - | 7 | 1047 | |||
Z48482.1 | - | 8 | 1372 |