Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197996.2 | NP_932113.2 | 4 | 542 | ||
GenBank mRNA | AK012471.1 | - | 4 | 541 | ||
AK172169.1 | - | 4 | 552 | |||
BC003872.1 | - | 4 | 491 | 3492667 | ||
CN455615.1 | - | 4 | 431 | |||
CX206586.1 | - | 4 | 542 |