Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082940.1 | - | 10 | 1001 | ||
AK167383.1 | - | 10 | 989 | |||
AK167848.1 | - | 10 | 985 | |||
AY082332.1 | - | 10 | 955 | |||
BC050206.1 | - | 10 | 963 | 30009463 |