Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005941.4 | NP_005932.2 | 7 | 1416 | ||
GenBank mRNA | AB009303.1 | - | 7 | 1246 | ||
AK314274.1 | - | 7 | 1416 | |||
AL832595.1 | - | 8 | 1455 | |||
BC069500.1 | - | 7 | 1203 | 7262415 | ||
BC075004.2 | - | 7 | 1160 | 30915400 | ||
BC075005.2 | - | 7 | 1160 | 30915613 | ||
D83646.1 | - | 7 | 1134 | |||
D83647.1 | - | 7 | 1134 | |||
D85511.1 | - | 7 | 1169 |