Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027427.2 | NP_081703.1 | 7 | 585 | ||
GenBank mRNA | AK011843.1 | - | 7 | 607 | ||
AK051568.1 | - | 7 | 586 | |||
AK078205.1 | - | 7 | 597 | |||
AK146331.1 | - | 7 | 548 | |||
AK166550.1 | - | 7 | 578 | |||
BC137591.1 | - | 7 | 559 | 8860610 |