Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011942.1 | - | 3 | 611 | ||
AK150539.1 | - | 3 | 729 | |||
AK151163.1 | - | 3 | 729 | |||
AK154611.1 | - | 3 | 680 | |||
BC020064.1 | - | 3 | 635 | |||
BC060191.1 | - | 3 | 604 | |||
BC080823.1 | - | 3 | 556 | |||
BC116944.1 | - | 3 | 654 | 40126263 | ||
BC116970.1 | - | 3 | 654 | 40126289 |