Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027432.3 | NP_081708.1 | 3, 4 | 526 | ||
GenBank mRNA | AK012014.1 | - | 3, 4 | 517 | ||
AK032897.1 | - | 1, 2 | 65 | |||
AK048488.1 | - | 3, 4 | 549 | |||
AK050391.1 | - | 3, 4 | 526 | |||
AK078856.1 | - | 4, 5 | 485 | |||
AK131608.1 | - | 2, 3 | 437 | |||
AK161812.1 | - | 3, 4 | 501 | |||
AK163839.1 | - | 521 | ||||
AK168006.1 | - | 3, 4 | 528 | |||
AK168752.1 | - | 3, 4 | 548 | |||
AK169128.1 | - | 3, 4 | 527 | |||
BC005755.1 | - | 3, 4 | 481 | 2654151 |