Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016155.5 | NP_057239.4 | 8 | 1294 | ||
XM_011538355.2 | XP_011536657.1 | 8 | 1148 | |||
XM_011538356.2 | XP_011536658.1 | 8 | 1144 | |||
XM_011538357.2 | XP_011536659.1 | 8 | 1093 | |||
XM_017019307.1 | XP_016874796.1 | 8 | 1273 | |||
GenBank mRNA | AB021225.2 | - | 8 | 1300 | ||
BC040507.1 | - | 8 | 1286 | 5301089 | ||
BC045610.1 | - | 8 | 1303 | 5273795 | ||
X89576.1 | - | 7 | 1172 |