Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001321.3 | NP_001001321.2 | 6 | 419 | ||
GenBank mRNA | AB117931.1 | - | 6 | 400 | ||
AB117932.1 | - | 6 | 400 | |||
AK154257.1 | - | 6 | 445 | |||
AK159148.1 | - | 6 | 420 | |||
BC107210.1 | - | 5 | 366 | |||
BC107211.1 | - | 6 | 399 | 40051417 |